missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Mouse anti-Human, Mouse, Pig, Rat, CoraLite 555, Clone: 2D7A9, Proteintech
Mouse Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | Calretinin |
|---|---|
| Clone | 2D7A9 |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | CoraLite 555 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17781359
|
Proteintech
CL555-66496-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17659906
|
Proteintech
CL555-66496-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calretinin Monoclonal antibody apecifically detetects Calretinin antigen in Human, Mouse, Pig, Rat Samples. It is validated for Immunofluorescence.Specifications
| Calretinin | |
| Immunofluorescence | |
| CoraLite 555 | |
| Mouse | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 794 | |
| Calretinin fusion protein Ag2924 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
| 2D7A9 | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Pig, Rat | |
| BC015484 | |
| CALB2 | |
| IgG1 | |
| Protein G purification |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title