Learn More
Calretinin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-32244
This item is not returnable.
View return policy
Description
Calretinin Polyclonal specifically detects Calretinin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Calretinin | |
| Western Blot 1:500-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen 1:100-1:1000 | |
| Rabbit | |
| 32 kDa |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| CALB2 | |
| Recombinant protein encompassing a sequence within the center region of human Calretinin. The exact sequence is proprietary. | |
| Affinity Purified |