Learn More
Calretinin Antibody (CL12842) [DyLight 755], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18545IR
Product Code. 30288731
  • £616.00 / 0.10mL
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Calretinin
Monoclonal
DyLight 755
29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR
This antibody was developed using a recombinant protein derived from P22676, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cellular Markers, Phospho Specific
794
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL12842
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Mouse
Purified