missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calretinin Antibody (CL12838), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18561-100ul
This item is not returnable.
View return policy
Description
Calretinin Monoclonal antibody specifically detects Calretinin in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Calretinin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL12838 | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| 29 kDa calbindin, CAB29, CAL2, calbindin 2, calbindin 2, (29kD, calretinin), calbindin 2, 29kDa (calretinin), calbindin D29K, calretinin, CR | |
| This antibody was developed against a recombinant protein of P22676. Actual immunogen sequence is proprietary. | |
| 100 μg | |
| Cellular Markers, Phospho Specific | |
| 794 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction