missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calreticulin Recombinant Rabbit Monoclonal Antibody (SAIC-16D-6B9)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA547895
This item is not returnable.
View return policy
Description
Specificity: Recognises human CALR.
Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
Specifications
| Calreticulin | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P27797 | |
| CALR | |
| Peptide GLQTSQDAR, derived from the calcium-binding protein chaperone Calreticulin, conjugated to KLH. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG κ |
| ELISA, Flow Cytometry, Western Blot, Immunocytochemistry, Mass Spectrometry | |
| SAIC-16D-6B9 | |
| Unconjugated | |
| CALR | |
| Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA | |
| Rabbit | |
| Protein A | |
| RUO | |
| 811 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction