missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calreticulin Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA3900

Product Code. 11518861

  • £455.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PA3-900 detects calreticulin from human, non-human primate, rabbit, mouse, canine and rat samples. PA3-900 has been successfully used in Western blot, immunofluorescence, immunocytochemistry, flow cytometry, and immunoprecipitation procedures. By Western blot, this antibody detects an ~60 kDa protein representing calreticulin from rat brain extract. Immunofluorescence staining of calreticulin in rat brain cortex with PA3-900 results in fine granular staining consistent with ER localization. PA3-900 immunogen is recombinant human calreticulin protein produced in the Baculovirus insect cell system.

Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calreticulin
Polyclonal
Unconjugated
CALR
Autoantigen RO; CABP3; CALBP; calcium-binding protein 3; CALR; calrectulin; calregulin; Calreticulin; calreticulin precursor; cC1qR; CRP55; CRT; CRTC; Endoplasmic reticulum resident protein 60; epididymis secretory sperm binding protein Li 99n; ERp60; FLJ26680; grp60; HACBP; HEL-S-99n; I79_008346; RO; Ro/SS-A Antigen; Sicca syndrome antigen A (autoantigen Ro; calreticulin); SSA
Rabbit
100 μL
Primary
Human, Mouse, Rat, Canine, Rabbit, Monkey
Antibody
IgG
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Conc. Not Determined
Whole serum with 0.05% sodium azide
P14211, P15253, P18418, P27797, P28490
CALR
Recombinant, human calreticulin protein.
RUO
100009050, 100350417, 12317, 476694, 64202, 811
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.