missing translation for 'onlineSavingsMsg'
Learn More

Calreticulin Antibody (CR213-2AG), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18395403 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395403 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395403 Supplier Novus Biologicals Supplier No. NBP311672AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calreticulin Monoclonal specifically detects Calreticulin in Human samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calreticulin
Applications Western Blot, ELISA
Classification Monoclonal
Clone CR213-2AG
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA
Host Species Mouse
Immunogen Recombinant human calreticulin.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 811
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.