missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calreticulin Antibody (681233) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB38981JF635
This item is not returnable.
View return policy
Description
Calreticulin Monoclonal antibody specifically detects Calreticulin in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Flow Cytometry (Intracellular Staining),Gene Knock-Out
Specifications
| Calreticulin | |
| Monoclonal | |
| Janelia Fluor 635 | |
| autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA | |
| E. coli-derived recombinant human Calreticulin, Glu18-Leu417, Accession # P27797 | |
| 0.1 mL | |
| Autophagy, Cancer, Cellular Markers, ER Markers, Immunology, Membrane Trafficking and Chaperones, Neurodegeneration, Neuroscience, Signal Transduction | |
| 811 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Immunohistochemistry, Western Blot, Flow Cytometry (Intracellular Staining), Gene Knock-Out | |
| 681233 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction