missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calreticulin Antibody (1G6A7), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Calreticulin Monoclonal antibody specifically detects Calreticulin in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
Specifications
| Antigen | Calreticulin |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1G6A7 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry, Flow (Intracellular) |
| Formulation | 50mM Sodium Borate |
| Gene Alias | autoantigen Ro, CALR, calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA |
| Host Species | Mouse |
| Immunogen | Calreticulin Antibody (1G6A7) was developed against a synthetic peptide corresponding to the C-terminus (EEEDVPGQAKDELC) of human Calreticulin, conjugated to KLH. [UniProt# P27797] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?