missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Calponin Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ Calponin Ab-1, Mouse Monoclonal Antibody.
£112.00 - £349.10
Specifications
Antigen | Calponin Ab-1 |
---|---|
Clone | CALP |
Applications | Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Concentration | Quantity | Price | Quantity & Availability | |||
12669176
|
Epredia™
MS-1168-P0 |
Concentrated | 200μg/mL | 100 μL |
£112.00
100µL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12679176
|
Epredia™
MS-1168-P1 |
Concentrated | 200μg/mL | 500 μL |
£303.00
500µL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12619186
|
Epredia™
MS-1168-R7 |
Prediluted | — | 7 mL |
£220.00
7mL |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12689176
|
Epredia™
MS-1168-P1ABX |
Concentrated | 1mg/mL | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12699176
|
Epredia™
MS-1168-PABX |
Concentrated | 1mg/mL | 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12659176
|
Epredia™
MS-1168-P |
Concentrated | 200μg/mL | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
12639186
|
Epredia™
MS-1168-RQ |
Ready-to-Use | — | 12 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Calponin, a calmodulin, binds tropomyosin and F-actin and is thought to be involved in the regulation of smooth muscle contraction. The expression of calponin is restricted to smooth muscle cells and is a marker of the differentiated contractile phenotype of developing smooth muscle. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. Expression of the 29 kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and malignant breast lesions.Host Species: Mouse
Clone: CALP
Isotype: IgG1/κ
Species Reactivity: Human and Rat. Others not tested.
Epitope: Not determined
Immunogen: Crude human uterus extract
Molecular Weight: 34 kDa
Positive Control: Myoepithelial cells in breast ducts
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Calponin Ab-1 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
IVD | |
Crude human uterus extract |
CALP | |
Monoclonal | |
Mouse | |
Muscle Biology | |
Primary |