missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calponin 3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584057
This item is not returnable.
View return policy
Description
Immunogen sequence: PKYCAAPTEP VIHNGSQGTG TNGSEISDSD YQAEYPDEYH GEYQDDYPRD YQYSDQGIDY Highest antigen sequence indentity to the following orthologs: Rat - 92%, Mouse - 93%.
This gene encodes a protein with a markedly acidic C terminus; the basic N-terminus is highly homologous to the N-terminus of a related gene, CNN1. Members of the CNN gene family all contain similar tandemly repeated motifs. This encoded protein is associated with the cytoskeleton but is not involved in contraction.
Specifications
| Calponin 3 | |
| Polyclonal | |
| Unconjugated | |
| CNN3 | |
| 1600014M03Rik; acidic calponin h3; C85854; Calpo3; calponin 3; calponin 3, acidic; calponin, acidic isoform; Calponin, non-muscle isoform; calponin-3; Cnn3; dJ639P13.2.2 (acidic calponin 3) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1266 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15417 | |
| CNN3 | |
| Recombinant protein corresponding to Human CNN3. Recombinant protein control fragment (Product #RP-103092). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction