missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calponin 2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549995
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total Calponin 2 protein.
The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| Calponin 2 | |
| Polyclonal | |
| Unconjugated | |
| Cnn2 | |
| AA408047; AI324678; Calpo2; calponin 2; calponin H2, smooth muscle; Calponin-2; CLP2; CNN2; h2-calponin; LOW QUALITY PROTEIN: calponin-2; neutral calponin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1265, 12798 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q08093, Q99439 | |
| Cnn2 | |
| Synthesized peptide derived from internal of human CNN2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction