Learn More
Invitrogen™ Calponin 2 Monoclonal Antibody (CP-93)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549306
Description
Adding 1 mL of PBS buffer will yield a concentration of 100 μg/mL. Positive Control - WB: chicken heart tissue, chicken heart tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene.
Specifications
| Calponin 2 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide | |
| Q99439 | |
| Cnn2 | |
| Turkey gizzard calponin. | |
| RUO | |
| 100216000, 1265 | |
| -20°C | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| CP-93 | |
| Unconjugated | |
| Cnn2 | |
| AA408047; AI324678; Calpo2; calponin 2; calponin H2, smooth muscle; Calponin-2; CLP2; CNN2; h2-calponin; LOW QUALITY PROTEIN: calponin-2; neutral calponin | |
| Mouse | |
| 100 μg | |
| Primary | |
| Avian, Chicken, Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.