missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin 2 Antibody (OTI2B5), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70421AF647
This item is not returnable.
View return policy
Description
Calponin 2 Monoclonal specifically detects Calponin 2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Calponin 2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CNN2 | |
| Full length recombinant protein of human Calponin 2 (NP_004359) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI2B5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| calponin 2, Calponin H2, smooth muscle, calponin-2, Neutral calponin | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 1265.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction