missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin 1 Rabbit anti-Human, Clone: CNN1/4227R, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-07589-20UG
This item is not returnable.
View return policy
Description
Calponin 1 Monoclonal specifically detects Calponin 1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
| Calponin 1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1 | |
| Synthetic peptide corresponding to residues on the C-terminus of human Calponin 1 was used as an immunogen. | |
| 20 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 1264 | |
| Store at 4°C. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CNN1/4227R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction