missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin 1 Antibody (CNN1/1408R), Novus Biologicals™
Rabbit Monoclonal Antibody
£227.00 - £432.00
Specifications
| Antigen | Calponin 1 |
|---|---|
| Clone | CNN1/1408R |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18217840
|
Novus Biologicals
NBP2-53233-20UG |
20 μg |
£227.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18203180
|
Novus Biologicals
NBP2-53233-100UG |
100 μg |
£432.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calponin 1 Monoclonal specifically detects Calponin 1 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Calponin 1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cytoskeleton Markers | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 1264 | |
| Recombinant human full-length CNN1 protein | |
| Primary | |
| Store at 4C. |
| CNN1/1408R | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1 | |
| CNN1 | |
| IgG κ | |
| Protein A or G purified | |
| 34 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title