missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin 1 Antibody (1H5B5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£161.00 - £435.00
Specifications
| Antigen | Calponin 1 |
|---|---|
| Clone | 1H5B5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:10-1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18237490
|
Novus Biologicals
NBP2-52445 |
0.1 mg |
£435.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18643837
|
Novus Biologicals
NBP2-52445-0.025mg |
0.025 mg |
£161.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calponin 1 Monoclonal specifically detects Calponin 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Calponin 1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cytoskeleton Markers | |
| PBS with 0.05% Sodium Azide | |
| 1264 | |
| Purified recombinant fragment of human CNN1 (AA: 16-165) expressed in E. Coli. | |
| Primary | |
| 33.2 kDa |
| 1H5B5 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Basic calponin, calponin 1, basic, smooth muscle, Calponin H1, smooth muscle, calponins, basic, Sm-Calp, SMCCcalponin-1 | |
| CNN1 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title