missing translation for 'onlineSavingsMsg'
Learn More

CalPlus™ (Brand of Calcium Chloride Dihydrate), Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15537904 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle
Unit Size:
2.5kg
500g
This item is not returnable. View return policy

Product Code. 15537904

Brand: Avantor J.T.Baker 133605

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for Use in Hemodialysis

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O

Specifications

Quantity 2.5 kg
Formula Weight 147.01
Grade Multi-Compendial/USP
Packaging Wide Mouth PVC Coated Glass Bottle
Specific Gravity 1L = 0.84kg
Chemical Name or Material CalPlus™ (Brand of Calcium Chloride Dihydrate)
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.