missing translation for 'onlineSavingsMsg'
Learn More

CalPlus™ (Brand of Calcium Chloride Dihydrate), Multi-Compendial, U.S.P., J.T. Baker™

Product Code. 15579400 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Wide Mouth PVC Coated Glass Bottle
Unit Size:
2.5kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15579400 500 g Wide Mouth Amber Glass Bottle 500g
15537904 2.5 kg Wide Mouth PVC Coated Glass Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15579400 Supplier Avantor J.T.Baker Supplier No. 133601

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for Use in Hemodialysis

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O

Specifications

Quantity 500 g
Formula Weight 147.01
Grade Multi-Compendial/USP
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 0.84kg
Chemical Name or Material CalPlus™ (Brand of Calcium Chloride Dihydrate)
compliance-icons
Product Identifier
  • Calcium Chloride Dihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Calcium Chloride Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.