missing translation for 'onlineSavingsMsg'
Learn More

Calpeptin, MedChemExpress

Product Code. 16499506
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499506 50 mg 50mg
16460906 200 mg 200mg
16455597 5 mg 5mg
16454266 100 mg 100mg
16432858 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16499506 Supplier Medchem Express Supplier No. HY100223
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Calpeptin is a potent, cell penetrating calpain inhibitor, with an ID50 of 40 nM for Calpain I in human platelets. Calpeptin is also an inhibitor of cathepsin K.

Calpeptin is a potent, cell penetrating calpain inhibitor, with an ID50 of 40 nM for Calpain I in human platelets. Calpeptin is also an inhibitor of cathepsin K.

Specifications

Chemical Name or Material Calpeptin
Physical Form Solid
CAS 117591-20-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H30N2O4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 100 mg/mL (275.89 mM; Need ultrasonic)
SMILES O=C(OCC1=CC=CC=C1)N[C@H](C(N[C@H](C=O)CCCC)=O)CC(C)C
Molecular Weight (g/mol) 362.46
Formula Weight 362.46
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.