missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpastatin Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12250-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an endogenous calpain (calcium-dependent cysteine protease) inhibitor. It consists of an N-terminal domain L and four repetitive calpain-inhibition domains (domains 1-4), and it is involved in the proteolysis of amyloid precursor protein. The calpain/calpastatin system is involved in numerous membrane fusion events, such as neural vesicle exocytosis and platelet and red-cell aggregation. The encoded protein is also thought to affect the expression levels of genes encoding structural or regulatory proteins. Alternatively spliced transcript variants encoding different isoforms have been described.Specifications
| Calpastatin | |
| Polyclonal | |
| Unconjugated | |
| CAST | |
| BS 17, Calpain inhibitor, calpastatin, CAST, Sperm BS 17 component | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 831 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P20810 | |
| CAST | |
| Calpastatin Fusion Protein Ag2897 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction