missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpastatin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583356
This item is not returnable.
View return policy
Description
Immunogen sequence: GPQNASSLKF EDAKLAAAIS EVVSQTPAST TQAGAPPRDT SQSDKDLDDA LDKLSDSLGQ RQPDPDENKP MEDKVKE Highest antigen sequence indentity to the following orthologs: Mouse - 75%, Rat - 69%.
Calpastatin is an endogenous calpain (calcium-dependent cysteine protease) inhibitor. It consists of an N-terminal domain L and four repetitive calpain-inhibition domains (domains 1-4), and it is involved in the proteolysis of amyloid precursor protein. The calpain/calpastatin system is involved in numerous membrane fusion events, such as neural vesicle exocytosis and platelet and red-cell aggregation. The protein is also thought to affect the expression levels of genes encoding structural or regulatory proteins.
Specifications
| Calpastatin | |
| Polyclonal | |
| Unconjugated | |
| Cast | |
| BS-17; Calpain inhibitor; calpastatin; calpastatin isoform III; calpastatin precursor (EC 3.4.22.17); calpastatin type 1; calpastatin type 2; calpastatin type 3; calpastatin type I; calpastatin type II; calpastatin type III; calpastatin type IV; Cast; Erythrocyte calpastatin; heart calpastatin; PLACK; Sperm BS-17 component; type III calpastatin | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 831 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20810 | |
| Cast | |
| Recombinant protein corresponding to Human CAST. Recombinant protein control fragment (Product #RP-102751). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction