missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpastatin Mouse anti-Human, Clone: CAST/1550, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | Calpastatin |
|---|---|
| Clone | CAST/1550 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA 2 to 4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
Description
Calpastatin Monoclonal specifically detects Calpastatin in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.Specifications
| Calpastatin | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Hypoxia, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| 831 | |
| IgG1 κ | |
| Protein A or G purified |
| CAST/1550 | |
| ELISA 2 to 4 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component | |
| Recombinant full-length human Calpastatin protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title