missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpastatin Monoclonal Antibody (A6B1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA544975
This item is not returnable.
View return policy
Description
Calpastatin is an endogenous calpain (calcium-dependent cysteine protease) inhibitor. It consists of an N-terminal domain L and four repetitive calpain-inhibition domains (domains 1-4), and it is involved in the proteolysis of amyloid precursor protein. The calpain/calpastatin system is involved in numerous membrane fusion events, such as neural vesicle exocytosis and platelet and red-cell aggregation. The protein is also thought to affect the expression levels of genes encoding structural or regulatory proteins.Specifications
Calpastatin | |
Monoclonal | |
2 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
P20810 | |
Cast | |
Recombinant protein within human Calpastatin aa 501-708/708. | |
100 μL | |
Primary | |
Human | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
A6B1 | |
Unconjugated | |
Cast | |
BS-17; Calpain inhibitor; calpastatin; calpastatin isoform III; calpastatin precursor (EC 3.4.22.17); calpastatin type 1; calpastatin type 2; calpastatin type 3; calpastatin type I; calpastatin type II; calpastatin type III; calpastatin type IV; Cast; Erythrocyte calpastatin; heart calpastatin; PLACK; Sperm BS-17 component; type III calpastatin | |
Mouse | |
Protein G | |
RUO | |
831 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |