missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpastatin Antibody (OTI1E7), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70338F
This item is not returnable.
View return policy
Description
Calpastatin Monoclonal specifically detects Calpastatin in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| Calpastatin | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| CAST | |
| Full length recombinant protein of human Calpastatin (NP_001741) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| OTI1E7 | |
| Flow Cytometry, Immunohistochemistry | |
| BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component | |
| Mouse | |
| Affinity Purified | |
| Hypoxia, Neuroscience | |
| 831.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction