missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpastatin Antibody (CAST/1550), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08510JF549
This item is not returnable.
View return policy
Description
Calpastatin Monoclonal antibody specifically detects Calpastatin in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Calpastatin | |
| Monoclonal | |
| Janelia Fluor 549 | |
| BS-17, Calpain inhibitor, calpastatin, MGC9402, Sperm BS-17 component | |
| Recombinant full-length human Calpastatin protein (Uniprot: P20810) | |
| 100 μg | |
| Hypoxia, Neuroscience | |
| 831 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| CAST/1550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction