missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain S2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62708-25ul
This item is not returnable.
View return policy
Description
Calpain S2 Polyclonal antibody specifically detects Calpain S2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Calpain S2 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Calcium-dependent protease small subunit 2, calpain small subunit 2, calpain, small subunit 2, CSS2, MGC12536, MGC14804 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KWQCVYKQYDRDHSGSLGSSQLRGALQAAGFQLNEQLYQMIVRRYANEDGD | |
| 25 μL | |
| Cell Biology, Neuroscience, Signal Transduction | |
| 84290 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction