missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calpain S1/S2 Monoclonal Antibody (28F3)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA112433

Product Code. 11588113

  • £455.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody specifically recognizes the 28 to 30 kD small subunit of Calpain 1 and Calpain 2 (mu calpain and m calpain). It recognizes both native and denatured protein in ELISA, Western Blot and Immunoprecipitation.

The calpains are calcium-dependent cysteine proteases that are widely expressed in mammalian systems. Both m-calpain (calpain II) and m-calpain (calpain I) are composed of an 80 kD subunit and a 30 kD subunit. Whereas the 30 kDa subunit is shared by both enzymes, the larger catalytic subunits are different and exhibit the distinct Ca++ requirements that are suggested by their names. Whereas m-calpain requires millimolar (mM) levels of calcium, m-calpain is active at micromolar (mM) concentrations of Ca++. In addition to the ubiquitously expressed m- and m- calpains, some tissue-specific calpains have been identified. The calpains appear to serve multiple physiological roles, and ideas concerning the functions of these enzymes are in a state of rapid flux.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calpain S1/S2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 3mM sodium azide; pH 7.5
P04632, P13135, Q96L46
CAPNS1, CAPNS2
Human Calpain protein purified from placenta.
100 μg
Primary
Human, Bovine
Antibody
IgG1
ELISA, Immunoprecipitation, Western Blot
28F3
Unconjugated
CAPNS1
2310005G05Rik; 30K-2; calcium-activated neutral proteinase 2; Calcium-activated neutral proteinase small subunit; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; calcium-dependent protease small subunit 2; calcium-dependent protease, small subunit; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain 4; calpain 4, small subunit (30K); calpain II regulatory subunit; calpain II regulatory subunit (EC 3.4.22.17); calpain large polypeptide L2; Calpain M-type; calpain regulatory subunit; Calpain small subunit 1; calpain small subunit 2; calpain, large polypeptide L2; calpain, small polypeptide; calpain, small subunit 1; calpain, small subunit 2; calpain-2 catalytic subunit; Calpain-2 large subunit; CALPAIN4; CANP; CANP 2; CANP small subunit; CANP2; CANPL2; CANPml; CANPS; Capa4; Capa-4; CAPN2; Capn4; CAPNS; CAPNS1; Capns2; CDPS; CSS1; CSS2; D7Ertd146e; M-calpain; mCANP; Millimolar-calpain
Mouse
Protein A/G
RUO
100300424, 281664, 826, 84290
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.