missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain S1 Monoclonal Antibody (2190CT339.12.2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA537795
This item is not returnable.
View return policy
Description
Calpain S1 Monoclonal Antibody for Western Blot
Calpains are a ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. Calpain families have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. Calpain I and II are heterodimeric with distinct large subunits associated with common small subunits, all of which are encoded by different genes. Calpain S1 is a small subunit common to both calpain I and II and is associated with myotonic dystrophy.
Specifications
| Calpain S1 | |
| Monoclonal | |
| Unconjugated | |
| CAPNS1 | |
| 30K; Calcium-activated neutral proteinase small subunit; calcium-dependent protease; Calcium-dependent protease small subunit; Calcium-dependent protease small subunit 1; calcium-dependent protease, small subunit; calpain 4; calpain 4, small subunit (30K); calpain II regulatory subunit; calpain II regulatory subunit (EC 3.4.22.17); calpain regulatory subunit; Calpain small subunit 1; calpain, small polypeptide; calpain, small subunit 1; CALPAIN4; CANP; CANP small subunit; CANPS; Capa4; Capa-4; Capn4; CAPNS; CAPNS1; CDPS; CSS1; D7Ertd146e; small pol; unknown protein | |
| Mouse | |
| Protein G | |
| RUO | |
| 100009090, 100770160, 826 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 2190CT339.12.2 | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P04632, P06813 | |
| CAPNS1 | |
| KLH conjugated synthetic peptide between 1-323 amino acids from the human region of human CAPNS1. | |
| 200 μL | |
| Primary | |
| Human, Rabbit, Hamster | |
| Antibody | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction