missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain S1 Antibody (1D11), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59446AF405
This item is not returnable.
View return policy
Description
Calpain S1 Monoclonal antibody specifically detects Calpain S1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| Calpain S1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Calcium-activated neutral proteinase small subunit, Calcium-dependent protease small subunit 1, calcium-dependent protease, small subunit, calpain 4, small subunit (30K), Calpain regulatory subunit, calpain small subunit 1, calpain, small polypeptide, CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1, epididymis secretory sperm binding protein | |
| Recombinant human Calpain S1 (84-268aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 826 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 1D11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction