missing translation for 'onlineSavingsMsg'
Learn More

Calpain Inhibitor I, 96%, ACROS Organics™

Product Code. 10674272 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10674272 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10674272 Supplier Thermo Scientific Acros Supplier No. 328150100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Purity ≥95%
Assay 96%
CAS 110044-82-1
Color White
Format Powder
Molecular Weight (g/mol) 383.533
Product Type Calpain Inhibitor I
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.