missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain 9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554197
This item is not returnable.
View return policy
Description
Immunogen sequence: PGEYILIPST FEPHQEADFC LRIFSEKKAI TRDMDGNVDI DLPEPPKPTP PDQETEEEQR FRALFEQVAG EDMEVTAEEL EYVLNAVLQK KKDIKFKKLS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.
Calpains are ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. The calpain proteins are heterodimers consisting of an invariant small subunit and variable large subunits. The large subunit possesses a cysteine protease domain, and both subunits possess calcium-binding domains. Calpains have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. The protein encoded by this gene is expressed predominantly in stomach and small intestine and may have specialized functions in the digestive tract. This gene is thought to be associated with gastric cancer. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Calpain 9 | |
| Polyclonal | |
| Unconjugated | |
| CAPN9 | |
| 2200003B16Rik; calpain 9; calpain-9; Capn9; Digestive tract-specific calpain; GC36; NCL4; nCL-4; New calpain 4; novel calpain large subunit-4; Protein CG36 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10753 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14815 | |
| CAPN9 | |
| Recombinant protein corresponding to Human Calpain 9. Recombinant protein control fragment (Product #RP-91742). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction