missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain 9 Monoclonal Antibody (OTI3G10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525714
This item is not returnable.
View return policy
Description
Calpain 9 Monoclonal Antibody for Western Blot, IHC (P)
Calpains are ubiquitous, well-conserved family of calcium-dependent, cysteine proteases. The calpain proteins are heterodimers consisting of an invariant small subunit and variable large subunits. The large subunit possesses a cysteine protease domain, and both subunits possess calcium-binding domains. Calpains have been implicated in neurodegenerative processes, as their activation can be triggered by calcium influx and oxidative stress. The protein encoded by this gene is expressed predominantly in stomach and small intestine and may have specialized functions in the digestive tract. This gene is thought to be associated with gastric cancer. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| Calpain 9 | |
| Monoclonal | |
| 0.8 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14815, O35920 | |
| CAPN9 | |
| Full length human recombinant protein of CAPN9 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Rat, Canine, Monkey | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3G10 | |
| Unconjugated | |
| CAPN9 | |
| 2200003B16Rik; calpain 9; calpain-9; Capn9; Digestive tract-specific calpain; GC36; NCL4; nCL-4; New calpain 4; novel calpain large subunit-4; Protein CG36 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10753, 116694, 608044 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction