missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92127-0.02ml
This item is not returnable.
View return policy
Description
Calpain 9 Polyclonal antibody specifically detects Calpain 9 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Calpain 9 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| calpain 9, calpain 9 (nCL-4), calpain-9, Digestive tract-specific calpain, EC 3.4.22, EC 3.4.22.54, GC36, NCL4, nCL-4EC 3.4.22.-, New calpain 4, novel calpain large subunit-4, Protein CG36 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 391-690 of human CAPN9 (NP_006606.1). QEECSFLVALMQKDRRKLKRFGANVLTIGYAIYECPDKDEHLNKDFFRYHASRARSKTFINLREVSDRFKLPPGEYILIPSTFEPHQEADFCLRIFSEKKAITRDMDGNVDIDLPEPPKPTPPDQETEEEQRFRALFEQVAGEDMEVTAEELEYVLNAVLQKKKDIKFKKLSLISCKNIISLMDTSGNGKLEFDEFKVFWDKLKQWINLFLRFDADKSGTMSTYELRTALKAAGFQLSSHLLQLIVLRYADEELQLDFDDFLNCLVRLENASRVFQALSTKNKEFIHLNINEFIHLTMNI | |
| 0.02 mL | |
| Cell Biology, Neuroscience, Signal Transduction | |
| 10753 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering