missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 9 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92127-0.1ml
This item is not returnable.
View return policy
Description
Calpain 9 Polyclonal antibody specifically detects Calpain 9 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Calpain 9 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| calpain 9, calpain 9 (nCL-4), calpain-9, Digestive tract-specific calpain, EC 3.4.22, EC 3.4.22.54, GC36, NCL4, nCL-4EC 3.4.22.-, New calpain 4, novel calpain large subunit-4, Protein CG36 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 391-690 of human CAPN9 (NP_006606.1). QEECSFLVALMQKDRRKLKRFGANVLTIGYAIYECPDKDEHLNKDFFRYHASRARSKTFINLREVSDRFKLPPGEYILIPSTFEPHQEADFCLRIFSEKKAITRDMDGNVDIDLPEPPKPTPPDQETEEEQRFRALFEQVAGEDMEVTAEELEYVLNAVLQKKKDIKFKKLSLISCKNIISLMDTSGNGKLEFDEFKVFWDKLKQWINLFLRFDADKSGTMSTYELRTALKAAGFQLSSHLLQLIVLRYADEELQLDFDDFLNCLVRLENASRVFQALSTKNKEFIHLNINEFIHLTMNI | |
| 0.1 mL | |
| Cell Biology, Neuroscience, Signal Transduction | |
| 10753 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction