missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain 7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103694
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CAPN7.
Calpain proteinases are defined currently as papain-like neutral proteases that are calcium activated. Calpains 1 and 2 are composed of a large subunit, which is proteolytically active, and a small subunit (also called calpain-4), or enhancer protein, that is not proteolytically active. The other calpain proteins identified to date (calpain 5, 6, 8, 9, 10, 11, 12, 13, 14, 15) are not known to require a small subunit. Domains in the large subunit include the amino terminal domain-I, the proteinase domain-II, domain-III, and the EF-hand domain-IV (domain-T in calpains 5 & 6, domain-N for calpain-7). Calpain-7 domain-N does not have the standard calpain EF hand domain for binding calcium, making it unclear whether it is calcium regulated. In place of the EF hand calcium-binding domain IV, calpain-7 has a sequence most closely resembling domain-III, with unknown function. Calpain-7 is widely distributed, found in most tissues analyzed.
Specifications
| Calpain 7 | |
| Polyclonal | |
| Unconjugated | |
| CAPN7 | |
| AU022319; calpain 7; calpain like protease; CALPAIN7; calpain-7; Capn7; homolog of Aspergillus Nidulans PALB; palB homolog; PALBH | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12339, 23473, 306260 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9R1S8, Q9Y6W3 | |
| CAPN7 | |
| A synthesized peptide derived from human CAPN7(Accession Q9Y6W3), corresponding to amino acid residues I298-D348. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction