missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 7 Polyclonal Antibody

Calpain 7 Polyclonal Antibody
Brand: Thermo Scientific PA5116762
This item is not returnable.
View return policy
Description
Calpain 7 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| Calpain 7 | |
| Polyclonal | |
| Unconjugated | |
| CAPN7 | |
| AU022319; calpain 7; calpain like protease; CALPAIN7; calpain-7; Capn7; homolog of Aspergillus Nidulans PALB; palB homolog; PALBH | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 12339, 23473, 306260 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.98 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9R1S8, Q9Y6W3 | |
| CAPN7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 20-220 of human CAPN7 (NP_0551111). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction