Learn More
Invitrogen™ Calpain 6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589430
Description
Immunogen sequence: MGPPLKLFKN QKYQELKQEC IKDSRLFCDP TFLPENDSLF YNRLLPGKVV WKRPQDICDD PHLIVGNISN HQLTQGRLGH KPMVSAFSCL AVQESHWTKT IPNHKEQEWD PQKTEKYAGI FHFRFWHFGE WTEVVIDDLL PTINGDLVFS FSTSMNEFWN ALLEKAYAKL LGCYEALDGL TITDIIVDFT GTLAETVDMQ KGRYTELVEE KYKLFGELYK TFTKGGLICC SIESPNQEEQ EVETDWGLLK GHTYTMTDIR KIRLGERLVE; Positive Samples: Rat heart; Cellular Location: Cytoplasm, cytoskeleton, perinuclear region, spindle.
Calpain-6, also known as calpamodulin or calpain-X, is an intracellular, calcium-dependent cysteine protease. Calpain-6 has a much more tissue-specific expression in adults than the ubiquitous calpain-1 and calpain-2, and has thus far been found primarily in the placenta, although it is expressed embryonically in a number of tissues. The classical calpain family members consist of a common small subunit (calpain-4), and a large variable subunit, but it is not known if calpain-6 associates with a small subunit. Domains in the large subunit include the amino terminal domain-I, the proteinase domain-II, domain-III, and the EF-hand domain-IV (domain T in calpains 5 and 6). Located on the X chromosome, the calpain-6 sequence lacks the EF hand calcium-binding motif found in domain-IV of the classical calpains. In addition, the canonical active site Cys, His and Asn are modified to Lys, His, Asn in human and Lys, Tyr, Asn in mouse calpain-6, making it unlikely that calpain-6 is proteolytically active.
Specifications
| Calpain 6 | |
| Polyclonal | |
| Unconjugated | |
| CAPN6 | |
| calpain 6; calpain-6; Calpain-like protease X-linked; calpamodulin; CalpM; CANPX; Capa6; CAPN6; CAPNX; DJ914P14.1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83685 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O88501 | |
| CAPN6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CAPN6 (NP_0551042). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.