missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Calpain 6 Monoclonal Antibody (1395CT446.22.5)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA524733

Product Code. 15994374

  • £441.00 / 400µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Calpain 6 Monoclonal Antibody for Western Blot, Flow

Calpain-6, also known as calpamodulin or calpain-X, is an intracellular, calcium-dependent cysteine protease. Calpain-6 has a much more tissue-specific expression in adults than the ubiquitous calpain-1 and calpain-2, and has thus far been found primarily in the placenta, although it is expressed embryonically in a number of tissues. The classical calpain family members consist of a common small subunit (calpain-4), and a large variable subunit, but it is not known if calpain-6 associates with a small subunit. Domains in the large subunit include the amino terminal domain-I, the proteinase domain-II, domain-III, and the EF-hand domain-IV (domain T in calpains 5 and 6). Located on the X chromosome, the calpain-6 sequence lacks the EF hand calcium-binding motif found in domain-IV of the classical calpains. In addition, the canonical active site Cys, His and Asn are modified to Lys, His, Asn in human and Lys, Tyr, Asn in mouse calpain-6, making it unlikely that calpain-6 is proteolytically active.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calpain 6
Monoclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9Y6Q1
CAPN6
human Calpain 6 recombinant protein.
400 μL
Primary
Human
Antibody
IgG1
Flow Cytometry, Western Blot
1395CT446.22.5
Unconjugated
CAPN6
calpain 6; calpain-6; Calpain-like protease X-linked; calpamodulin; CalpM; CANPX; Capa6; CAPN6; CAPNX; DJ914P14.1
Mouse
Protein G
RUO
827
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documenti

Documenti

Certificati
Promozioni

Promozioni

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato