missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92126-0.02ml
This item is not returnable.
View return policy
Description
Calpain 6 Polyclonal antibody specifically detects Calpain 6 in Rat samples. It is validated for Western Blot
Specifications
| Calpain 6 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| calpain 6, calpain-6, Calpain-like protease X-linked, Calpamodulin, CalpMCALPM, CANPXcalpamodulin, CAPNX, DJ914P14.1, EC 3.4.22 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CAPN6 (NP_055104.2). MGPPLKLFKNQKYQELKQECIKDSRLFCDPTFLPENDSLFYNRLLPGKVVWKRPQDICDDPHLIVGNISNHQLTQGRLGHKPMVSAFSCLAVQESHWTKTIPNHKEQEWDPQKTEKYAGIFHFRFWHFGEWTEVVIDDLLPTINGDLVFSFSTSMNEFWNALLEKAYAKLLGCYEALDGLTITDIIVDFTGTLAETVDMQKGRYTELVEEKYKLFGELYKTFTKGGLICCSIESPNQEEQEVETDWGLLKGHTYTMTDIRKIRLGERLVE | |
| 0.02 mL | |
| Cell Biology, Neuroscience, Signal Transduction | |
| 827 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction