missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain 5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521486
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3. Predicted reactivity: Mouse (93%), Rat (92%), Xenopus laevis (81%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Calpains are calcium-dependent cysteine proteases involved in signal transduction in a variety of cellular processes. A functional calpain protein consists of an invariant small subunit and 1 of a family of large subunits. CAPN5 is one of the large subunits. Unlike some of the calpains, CAPN5 and CAPN6 lack a calmodulin-like domain IV. Because of the significant similarity to Caenorhabditis elegans sex determination gene tra-3, CAPN5 is also called as HTRA3.
Specifications
| Calpain 5 | |
| Polyclonal | |
| Unconjugated | |
| CAPN5 | |
| ADNIV; calpain 5; calpain htra-3; Calpain-5; Capn5; high-temperature requirement factor A3; HTRA3; Ncl3; nCL-3; New calpain 3; testis tissue sperm-binding protein Li 91mP; VRNI | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12337, 726 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.47 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| O08688, O15484 | |
| CAPN5 | |
| Recombinant protein encompassing a sequence within the center region of human Calpain 5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction