missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain 2 Recombinant Rabbit Monoclonal Antibody (23GB3295), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553402
This item is not returnable.
View return policy
Description
Mu-Calpain, also known as Calpain-I, and m-calpain, also known as Calpain-II, are intracellular, calcium-dependent cysteine proteases. Mu-calpain has a micromolar sensitivity (thus the mu-) as compared to the millimolar calcium sensitivity of m-calpain.
Specifications
| Calpain 2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| CAPN2 | |
| 80 kDa M-calpain subunit; AI326419; Calcium-activated neutral proteinase 2; calcium-dependent protease; CALP80; calpain 2; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; calpain II; calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; CANP2; CANPL2; CANPml; Capa2; Capa-2; CAPN2; cysteine proteinase; m-calpain; m-calpain 80 kDa large subunit; m-calpain large subunit; m-calpin; mCANP; millimolar-calpain | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12334, 29154, 824 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB3295 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O08529, P17655, Q07009 | |
| CAPN2 | |
| A synthetic peptide of human Calpain 2 (657-700AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction