missing translation for 'onlineSavingsMsg'
Learn More

Calpain 2 Antibody (OTI1F4), DyLight 594, Novus Biologicals™

Product Code. 18413213 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413213 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413213 Supplier Novus Biologicals Supplier No. NBP270335DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calpain 2 Monoclonal antibody specifically detects Calpain 2 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 2
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1F4
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Calcium-activated neutral proteinase 2, calpain 2, (m/II) large subunit, calpain 2, large [catalytic] subunit, calpain 2, large subunit, Calpain large polypeptide L2, Calpain M-type, calpain, large polypeptide L2, calpain-2 catalytic subunit, Calpain-2 large subunit, CANP 2, CANPL2CANP2, CANPml, EC 3.4.22, EC 3.4.22.53, FLJ39928, M-calpain, mCANP, Millimolar-calpain
Host Species Mouse
Immunogen Full length recombinant protein of human Calpain 2 (NP_001739) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Cell Biology, Endocrinology, Immunology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 824
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.