missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 15 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32098AF750
This item is not returnable.
View return policy
Description
Calpain 15 Polyclonal antibody specifically detects Calpain 15 in Human samples. It is validated for Western Blot
Specifications
| Calpain 15 | |
| Polyclonal | |
| Western Blot | |
| calpain-15, CAPN15Small optic lobes homolog, EC 3.4.22, EC 3.4.22.-, EC 3.4.22.54, MGC131491, small optic lobes (Drosophila) homolog, small optic lobes homolog (Drosophila) | |
| The immunogen this antibody was made to, maps to a region between residue 25 to 75 of human Small Optic Lobes Homolog using the numbering given in entry NP_005623.1 (GeneID 6650). | |
| 0.1 mL | |
| Vision | |
| 6650 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction