missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13675-1-AP-20UL
This item is not returnable.
View return policy
Description
Calpains constitute a family of intracellular calcium-dependent cysteine proteases. There are eight members in this superfamily. They consist of a variable 80 kDa subunit and an invariant 30 kDa subunit. This calpain protein appears to have protease activity and calcium-binding ability. A similar mouse protein may play a functional role in spermatogenesis and in the regulation of calcium-dependent signal transduction events during meiosis.Specifications
| Calpain 11 | |
| Polyclonal | |
| Unconjugated | |
| CAPN11 | |
| Calpain 11, calpain11, CANP 11, CAPN11 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11131, 268958, 408218 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q4V8Q1, Q6J756, Q9UMQ6 | |
| Capn11 | |
| Calpain 11 Fusion Protein Ag4668 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction