missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calpain 10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106623
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total CAPN10.
The calpains including Calpain 1 (CAPN1), calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 1. Several transcript variants encoding two different isoforms have been found for this gene. Diseases associated with CAPN1 include Spastic Paraplegia 76, Autosomal Recessive and Spasticity.
Specifications
| Calpain 10 | |
| Polyclonal | |
| Unconjugated | |
| CAPN10 | |
| calcium-activated neutral proteinase 10; calpain 10; calpain-10; calpain-like protease CAPN10; CANP 10; CANP10; CAPN10; KIAA1845; NIDDM1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11132, 23830 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9ESK3, Q9HC96 | |
| CAPN10 | |
| A synthesized peptide derived from human CAPN10(Accession Q9HC96), corresponding to amino acid residues L34-K84. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction