missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Calpain 10 Polyclonal antibody specifically detects Calpain 10 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Calpain 10 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS (pH 7.4) |
| Gene Accession No. | NP_075571.1 |
| Gene Alias | Calcium-activated neutral proteinase 10, calpain 10, calpain-10, calpain-like protease CAPN10, CANP 10, CANP10, EC 3.4.22.-, KIAA1845, NIDDM1 |
| Host Species | Rabbit |
| Immunogen | CAPN10 (NP_075571.1, 1 a.a. - 672 a.a.) full-length human protein. MRAGRGATPARELFRDAAFPAADSSLFCDLSTPLAQFREDITWRRPQEICATPRLFPDDPREGQVKQGLLGDCWFLCACAALQKSRHLLDQVIPPGQPSWADQEYRGSFTCRIWQFGRWVEVTTDDRLPCLAGRLCFSRCQREDVFWLPLLEKVYAKVHGSYEHLWAGQVADALVDLTGGLAERWNLKGVAGSGGQQDRPGRWEHRTCRQLLHLKDQCLISCCVLSPRAGARELGEFHAFIVSDLRELQGQAGQCILLLRIQNPWGRRCWQGLWREGGEGWSQVDAAVASELLSQLQEGEFWVEEEEFLREFDELTVGYPVTEAGHLQSLYTERLLCHTRALPGAWVKGQSAGGCRNNSGFPSNPKFWLRVSEPSEVYIAVLQRSRLHAADWAGRARALVGDSHTSWSPASIPGKHYQAVGLHLWKVEKRRVNLPRVLSMPPVAGTACHAYDREVHLRCELSPGYYLAVPSTFLKDAPGEFLLRVFSTGRVSLSAIRAVAKNTTPGAALPAGEWGTVQLRGSWRVGQTAGGSRNFASYPTNPCFPFSVPEGPGPRCVRITLHQHCRPSDTEFHPIGFHIFQVPEGGRSQDAPPLLLQEPLLSCVPHRYAQEVSRLCLLPAGTYKVVPSTYLPDTEGAFTVTIATRIDRPSIHSQEMLGQFLQEVSVMAVMKT |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?