missing translation for 'onlineSavingsMsg'
Learn More

Calpain 1 Mouse anti-Human, Clone: CAPN1/1530, Novus Biologicals™

Product Code. 18392336 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392336 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392336 Supplier Novus Biologicals Supplier No. NBP30738720UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Calpain 1 Monoclonal specifically detects Calpain 1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calpain 1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CAPN1/1530
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution ELISA 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml
Formulation 10 mM PBS with 0.05% BSA
Gene Alias Calcium-activated neutral proteinase 1, calpain 1, (mu/I) large subunit, Calpain mu-type, calpain, large polypeptide L1, calpain-1 catalytic subunit, Calpain-1 large subunit, CANP, CANP 1, CANPL1EC 3.4.22.52, Cell proliferation-inducing gene 30 protein, cell proliferation-inducing protein 30, EC 3.4.22, Micromolar-calpain, muCANPCANP1, muCL
Host Species Mouse
Immunogen Recombinant full-length human Calpain 1 protein
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 823
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.