missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calpain 1 Antibody (CAPN1/1530), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08456V
This item is not returnable.
View return policy
Description
Calpain 1 Monoclonal antibody specifically detects Calpain 1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Calpain 1 | |
| Monoclonal | |
| DyLight 405 | |
| Calcium-activated neutral proteinase 1, calpain 1, (mu/I) large subunit, Calpain mu-type, calpain, large polypeptide L1, calpain-1 catalytic subunit, Calpain-1 large subunit, CANP, CANP 1, CANPL1EC 3.4.22.52, Cell proliferation-inducing gene 30 protein, cell proliferation-inducing protein 30, EC 3.4.22, Micromolar-calpain, muCANPCANP1, muCL | |
| Recombinant full-length human Calpain 1 protein (Uniprot: P07384) | |
| 100 μg | |
| Cell Cycle and Replication | |
| 823 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| CAPN1/1530 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction