missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Calnexin Recombinant Rabbit Monoclonal Antibody (23GB2760), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | Calnexin |
|---|---|
| Clone | 23GB2760 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088979
![]() |
Invitrogen™
MA553476 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089647
|
Invitrogen™
MA553477 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calnexin Recombinant Monoclonal antibody specifically detects Calnexin in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Calnexin | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P27824 | |
| 821 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2760 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110069N15Rik; AI988026; calmegin; calnexin; canx; canx-a; canx-b; clgn; clnx; cnx; D11Ertd153e; FLJ26570; hypothetical protein; IP90; Major histocompatibility complex class I antigen-binding protein p88; p88; P90; PP90; pp90 precursor | |
| A synthesized peptide derived from human Calnexin (550-592AA). | |
| Primary | |
| -20°C | |
| CANX |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title